Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TDRD1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6296
This item is not returnable.
View return policy
Description
TDRD1 Polyclonal specifically detects TDRD1 in Human samples. It is validated for Western Blot.Specifications
TDRD1 | |
Polyclonal | |
Unconjugated | |
Q9BXT4 | |
TDRD1 | |
E. coli-derived recombinant human TDRD1 Asn837-Lys968 Accession # Q9BXT4 | |
100 μg | |
Primary | |
Detects human TDRD1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse TDRD1 is observed and less than 1% cross-reactivity with recombinant human TDRKH is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
CT41.1, CT41.1Cancer/testis antigen 41.1, FLJ21082, tudor domain containing 1, tudor domain containing protein 1, tudor domain-containing protein 1 | |
Sheep | |
Affinity Purified | |
RUO | |
56165 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction