Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TEM5/GPR124 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8896N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8896N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8896N100 Supplier R&D Systems Supplier No. FAB8896N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TEM5/GPR124 Monoclonal specifically detects TEM5/GPR124 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TEM5/GPR124
Applications Flow Cytometry
Classification Monoclonal
Clone 499716
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q96PE1
Gene Alias DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, GPR124, G-protein coupled receptor 124, KIAA1531, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human TEM5/GPR124 Met1-Leu699 Accession # Q96PE1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25960
Test Specificity Detects human TEM5/GPR124 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.