Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TEM5/GPR124 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8896
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8896 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8896 Supplier R&D Systems Supplier No. MAB8896
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TEM5/GPR124 Monoclonal specifically detects TEM5/GPR124 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen TEM5/GPR124
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 499716
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96PE1
Gene Alias DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, GPR124, G-protein coupled receptor 124, KIAA1531, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5
Gene Symbols GPR124
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human TEM5/GPR124 Met1-Leu699 Accession # Q96PE1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25960
Test Specificity Detects human TEM5/GPR124 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.