Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TEM8/ANTXR1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3886
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3886 100 μg
MAB3886SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3886 Supplier R&D Systems Supplier No. MAB3886
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TEM8/ANTXR1 Monoclonal specifically detects TEM8/ANTXR1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TEM8/ANTXR1
Applications Western Blot
Classification Monoclonal
Clone 371113
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9H6X2
Gene Alias anthrax toxin receptor 1, ANTXR1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8
Gene Symbols ANTXR1
Host Species Mouse
Immunogen E. coli-derived recombinant human TEM8/ANTXR1 isoform 1 Gln28-Gly320 Accession # Q9H6X2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84168
Test Specificity Detects human TEM8/ANTXR1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.