Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tenascin X Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6999
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6999 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6999 Supplier R&D Systems Supplier No. MAB6999
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tenascin X Monoclonal specifically detects Tenascin X in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Tenascin X
Applications Western Blot
Classification Monoclonal
Clone 766842
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH33740
Gene Alias Hexabrachion-like protein, HXBL, tenascin XB, tenascin XB2, tenascin-X, TNX, TNXB, TNXB1growth-inhibiting protein 45, TNXB2, TNXB2tenascin XB1, TNXBS, TNXBSTNXTENX, XB, XBS, XB-S, XBTN-X
Gene Symbols TNXB
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Tenascin XB2 Met1-Tyr673 Accession # AAH33740
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7148
Test Specificity Detects human Tenascin XB2 in ELISAs and Western blots. Also detects Tenascin X in Western blots. In direct ELISAs, no cross-reactivity with recombinant human Tenascin R is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.