Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TFEB Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 2 publications

$214.00 - $538.00

Specifications

Antigen TFEB
Clone 954604
Dilution Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB9170SP
GREENER_CHOICE
View Documents
R&D Systems
MAB9170SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB9170100
GREENER_CHOICE
View Documents
R&D Systems
MAB9170100
100 μg
Each for $538.00
Only null left
Add to Cart
 
Description

Description

TFEB Monoclonal specifically detects TFEB in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications

Specifications

TFEB
Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEB, TCFEBbHLHe35, transcription factor EB
TFEB
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
954604
Western Blot, Immunocytochemistry
Unconjugated
Mouse
Human
P19484
7942
Human TFEB synthetic peptide Pro384-Ala446 Accession # P19484
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human TFEB in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.