Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TFPI-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | TFPI-2 |
---|---|
Clone | 243227 |
Dilution | Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL |
Applications | Immunoprecipitation, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB25451SP
![]() |
R&D Systems
MAB25451SP |
25 μg |
Each for $200.50
|
|
|||||
MAB25451
![]() |
R&D Systems
MAB25451 |
500 μg |
Each for $655.00
|
|
|||||
Description
TFPI-2 Monoclonal specifically detects TFPI-2 in Human samples. It is validated for Immunoprecipitation, Neutralization.Specifications
TFPI-2 | |
Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Placental protein 5, PP5, PP5FLJ21164, REF-1, TFPI2, TFPI-2REF1, tissue factor pathway inhibitor 2 | |
TFPI2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
243227 | |
Immunoprecipitation, Neutralization | |
Unconjugated | |
Mouse | |
Human | |
P48307 | |
7980 | |
Mouse myeloma cell line NS0-derived recombinant human TFPI-2 Asp23-Phe235 Accession # P48307 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TFPI-2 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title