Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TFPI Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB29741
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB29741 500 μg
MAB29741SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB29741 Supplier R&D Systems Supplier No. MAB29741
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TFPI Monoclonal specifically detects TFPI in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TFPI
Applications ELISA
Classification Monoclonal
Clone 374718
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10646
Gene Alias convertin, EPI, EPITFPI1LACITFI, Extrinsic pathway inhibitor, LACI, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Gene Symbols TFPI
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TFPI Asp29-Lys282 Accession # P10646
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7035
Test Specificity Detects human TFPI in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Coagulation factor II, VII, XI, XIV, rhTFPI-2, recombinant mouse (rm) TFPI, rmTFPI-2, or rhVitronectin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.