Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TfR (Transferrin R) Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2474V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2474V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2474V100 Supplier R&D Systems Supplier No. FAB2474V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TfR (Transferrin R)
Applications Flow Cytometry
Classification Monoclonal
Clone 29806
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Gene Symbols TFRC
Host Species Mouse
Immunogen Human TfR
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7037
Test Specificity Detects human TfR in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.