Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TfR2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3120100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3120100 100 μg
AF3120SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3120100 Supplier R&D Systems Supplier No. AF3120100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TfR2 Polyclonal specifically detects TfR2 in Human samples. It is validated for Western Blot.

Specifications

Antigen TfR2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 50 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9UP52
Gene Alias HFE3, HFE3transferrin receptor protein 2, MGC126368, TfR2, TFRC2, transferrin receptor 2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TfR2 Arg105-Phe801 Accession # Q9UP52
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7036
Test Specificity Detects human TfR2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.