Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TGF-beta 1 (ELISA Standard) Recombinant Protein

Catalog No. 501130698
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-0698 4 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-0698 Supplier Invitrogen™ Supplier No. 39834365
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Description: Transforming Growth Factor beta (TGF beta) is a pleiotropic cytokine which exists in five isoforms, known as TGF beta 1-5, with homologies of 70-80% and no homology to TGF alpha. TGF beta 1 is the most abundant isoform and is ubiquitously expressed, while other isoforms are expressed in a more restricted manner. TGF beta 1 is highly conserved, with 100% sequence homology between the human, simian, bovine, porcine, and chicken proteins and 99% homology between the human and murine proteins. It is highly expressed in platelets and also produced by macrophages, lymphocytes, endothelial cells, chondrocytes, and leukemic cells. TGF beta 1 is synthesized as a long precursor polypeptide, which is cleaved to yield the mature protein and the Latency Associated Peptide (LAP). LAP and mature TGF beta 1 remain non-covalently associated through secretion, forming homodimers known as the Small Latent Complex (SLC). Secretion can be induced by steroids, retinoids, EGF, NGF, vitamin D3, and IL-1. The bioactivity of mature TGF beta 1 is dependent on its release from LAP by conformational changes and proteolytic processing. Its activities include inhibition of cell growth in epithelial cells, endothelial cells, fibroblasts, neurons, lymphoid cells, and other hematopoietic cell types.

TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01137
For Use With (Application) Neutralization, ELISA Standard
Formulation Protein with 0.09% sodium azide
Gene ID (Entrez) 7040
Name Human TGF-beta 1 (ELISA Standard)
Purification Method Purified
Quantity 4 pack
Source CHO cells
Storage Requirements -80°C
Regulatory Status RUO
Gene Alias CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF β; TGF β 1; TGF β1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFβ1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor β1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product
Common Name TGF beta-1
Gene Symbol Tgfb1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage -80°C
Expression System CHO cells
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.