Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human/Mouse/Rat/Porcine/Canine TGF-beta 1 Quantikine ELISA
Click to view available options
Quantity:
6 Plates
Description
The Human TGF-beta 1 Quantikine ELISA Kit quantifies human TGF-beta 1 in cell culture supernates (100 ul), serum (40 ul), edta plasma (40 ul), urine (100 ul).
Specifications
Specifications
Assay Range | 31.20 - 2000pg/mL |
Assay Sensitivity | 15.4pg/mL |
Conjugate | HRP |
Cross Reactivity | Cross-species reactivity observed with 1 or more species tested. Cross-reactivity observed with 1 or more available related molecules |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Platelet-poor Plasma, Serum, Urine |
Sufficient For | 6 x 96-well Plates |
Target Species | Human |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 7040 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction