Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TGF-beta 2 Recombinant Protein, PeproTech®

Catalog No. p-7245948
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 250 μg
250 μg
4 x 250 μg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

100-35B-1MG will be provided as 4 x 250 μg (100-35B-250UG). 100-35B-500UG will be provided as 2 x 250 μg (100-35B-250UG). Recombinant Human TGF-beta2 is a 25.0 kDa protein composed of two identical 112 amino acid polypeptide chains linked by a single disulfide bond. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Transforming Growth Factor (TGF) betas mediate many cell to cell interactions that occur during embryonic development. Three TGF betas have been identified in mammals. TGF beta 1, TGF beta 2 and TGF beta 3 are each synthesized as precursor proteins that are very similar in that each is cleaved to yield a 112 amino acid polypeptide that remains associated with the latent portion of the molecule. The TGF beta polypeptides are multifunctional; capable of influencing cell proliferation, differentiation, and other functions in a wide range of cell types. Transformed, as well as nonneoplastic tissues, release transforming growth factors; and essentially all mammalian cells possess a specific TGF receptor. The multi modal nature of TGF beta is seen in its ability to stimulate or inhibit cellular proliferation. In general, cells of mesenchymal origin appear to be stimulated by TGF beta whereas cells of epithelial or neuroectodermal origin are inhibited by the peptide. TGF beta 1, TGF beta 2, and TGF beta 1.2 appear to be equivalent in biological activity, although there does appear to be differences in binding to certain types of receptors. TGF beta 2 is produced by many cell types and has been found in the highest concentration in porcine platelets and mammalian bone. Latent TGF beta 2 is the prominent isoform found in body fluids such as amniotic fluid, breast milk, and the aqueous and vitreous humor of the eye.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P61812
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7042
Molecular Weight (g/mol) 25 kDa
Name Human TGF-beta 2
Quantity 100 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BB105277; BSC-1 cell growth inhibitor; cetermin; glioblastoma-derived T-cell suppressor factor; G-TSF; LAP; Latency-associated peptide; LDS4; polyergin; prepro-transforming growth factor beta-2; TGF beta 2 protein; TGFB; TGFB2; TGF-B2; Tgfb-2; TGF-beta2; TGF-beta-2; TGFβ2; Transforming growth factor; transforming growth factor beta 2; transforming growth factor beta-2; Transforming growth factor beta-2 proprotein; transforming growth factor, beta 2
Common Name TGF beta-2
Gene Symbol TGFB2
Biological Activity Determined by its ability to inhibit the mouse IL-4-dependent proliferation of mouse HT-2 cells. The ED50 was found to be ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence ALDAAYCFRN VQDNCCLRPL YIDFKRDLGW KWIHEPKGYN ANFCAGACPY LWSSDTQHSR VLSLYNTINP EASASPCCVS QDLEPLTILY YIGKTPKIEQ LSNMIVKSCK CS
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.