Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TGF-beta RI/ALK-5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3025SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3025SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3025SP Supplier R&D Systems Supplier No. AF3025SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 7 publications

TGF-beta RI/ALK-5 Polyclonal specifically detects TGF-beta RI/ALK-5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry.

Specifications

Antigen TGF-beta RI/ALK-5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1-0.3 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P36897
Gene Alias AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I
Gene Symbols TGFBR1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TGF-beta RI/ALK-5 Leu34-Glu125 Accession # P36897
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7046
Test Specificity Detects human TGF-beta RI/ALK-5 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse TGF-beta RI is observed and less than 5% cross-reactivity with recombinant human (rh) TGF-beta RII and rhTGF-beta RIII is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.