Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGF-beta RII Isoform 2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $561.50
Specifications
Antigen | TGF-beta RII |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
TGF-beta RII Polyclonal specifically detects TGF-beta RII in Human samples. It is validated for Western Blot.Specifications
TGF-beta RII | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
TGFBR2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_001020018 | |
7048 | |
Mouse myeloma cell line NS0-derived recombinant human TGF-beta RII Isoform 2 Thr23-Asp184 Accession # NP_001020018 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human TGF-beta RII Isoform 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human (rh) TGF-beta RII, recombinant mouse (rm) TGF-beta RII, rmTGF-beta RI, and rhTGF-beta RIII is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title