Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGF-beta RII PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$268.50 - $514.50
Specifications
| Antigen | TGF-beta RII |
|---|---|
| Clone | 25508 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB241C025
![]() |
R&D Systems
FAB241C025 |
25 Tests |
Each for $268.50
|
|
|||||
FAB241C100
![]() |
R&D Systems
FAB241C100 |
100 Tests |
Each for $514.50
|
|
|||||
Description
TGF-beta RII Monoclonal specifically detects TGF-beta RII in Human samples. It is validated for Flow Cytometry.Specifications
| TGF-beta RII | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
| TGFBR2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| This antibody has been validated by flow cytometry on TGF-beta RII-transfected cells only. In contrast to Catalog # FAB2411A, minimal staining of PBMC lymphocytes was observed with this antibody. |
| 25508 | |
| Flow Cytometry | |
| PerCP | |
| Mouse | |
| Human | |
| P37173.2 | |
| 7048 | |
| Mouse myeloma cell line NS0-derived recombinant human TGF-beta RII Ile24-Asp159 Accession # P37173.2 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title