Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGF-beta RIII Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | TGF-beta RIII |
---|---|
Clone | 31308 |
Dilution | ELISA Capture (Matched Antibody Pair) 0.5-2.0 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
TGF-beta RIII Monoclonal specifically detects TGF-beta RIII in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
TGF-beta RIII | |
ELISA Capture (Matched Antibody Pair) 0.5-2.0 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
betaglycan, betaglycan proteoglycan, BGCAN, TBRIII, TGF-beta receptor type 3, TGF-beta receptor type III, TGFbetaRIII, TGFBR3, TGF-bRIII, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) | |
TGFBR3 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
31308 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
AAA67061 | |
7049 | |
Murine myeloma cell line, NS0-derived recombinant human TGF-beta RIII Gly21-Asp781 Accession # AAA67061 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TGF-beta RIII in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) TGF-beta RI, RII, RIIb, recombinant mouse (rm) TGF-beta RIII, rhTGF-beta 1, beta 1.2, beta 2, beta 3, rhLatent TGF-beta 1, or rhLAP is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title