Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGF-beta RIII Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF242
This item is not returnable.
View return policy
Description
TGF-beta RIII Polyclonal specifically detects TGF-beta RIII in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
TGF-beta RIII | |
Polyclonal | |
Biotin | |
AAA67061 | |
TGFBR3 | |
Mouse myeloma cell line NS0-derived recombinant human TGF-beta RIII Gly21-Asp781 Accession # AAA67061 | |
50 μg | |
Primary | |
Detects human TGF-beta RIII in ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) TGF-beta 1, rhTGF-beta 2, rhTGF-beta 3, recombinant amphibian TGF-beta 5, rhLAP (TGF-beta 1), rhTGF-beta RII, and rhGDNF is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-.04 ug/mL | |
betaglycan, betaglycan proteoglycan, BGCAN, TBRIII, TGF-beta receptor type 3, TGF-beta receptor type III, TGFbetaRIII, TGFBR3, TGF-bRIII, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) | |
Goat | |
Affinity Purified | |
RUO | |
7049 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction