Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGF-beta RIII PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems FAB242P
This item is not returnable.
View return policy
Description
TGF-beta RIII Polyclonal specifically detects TGF-beta RIII in Human samples. It is validated for Flow Cytometry.Specifications
TGF-beta RIII | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
betaglycan, betaglycan proteoglycan, BGCAN, TBRIII, TGF-beta receptor type 3, TGF-beta receptor type III, TGFbetaRIII, TGFBR3, TGF-bRIII, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) | |
Goat | |
Affinity Purified | |
RUO | |
7049 | |
Human | |
IgG |
Flow Cytometry | |
R-PE | |
AAA67061 | |
TGFBR3 | |
Mouse myeloma cell line NS0-derived recombinant human TGF-beta RIII Gly21-Asp781 Accession # AAA67061 | |
100 Tests | |
Primary | |
Detects human TGF-beta RIII in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) TGF-beta RIII is observed, and less than 1% cross-reactivity with recombinant human (rh) TGF-beta RI, rhTGF-beta RII, rhTGF-beta RIIB, and rmTGF-beta RI is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction