Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TGFB2 (P61812, 112 amino acids) Partial Recombinant Protein

Catalog No. 89964542
Click to view available options
Quantity:
5 μg

Used for Func, SDS-PAGE

Specifications

Accession Number P61812
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 7042
Molecular Weight (g/mol) 25kDa
Name TGFB2 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 5 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with 1 mM citrate buffer, pH 3.5, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias MGC116892/TGF-beta2
Common Name TGFB2
Gene Symbol TGFB2
Biological Activity The ED50 as determined by the ability of TGF-beta2 to inhibit the mouse IL-4-dependent proliferation of mouse HT-2 cells is < 0.2ng/mL, corresponding to a specific activity of > 5 x 106 units/mg.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥98% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.