Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGIF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7555SP
This item is not returnable.
View return policy
Description
TGIF1 Monoclonal specifically detects TGIF1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
| TGIF1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL | |
| Q15583 | |
| TGIF1 | |
| E. coli-derived recombinant human TGIF1 Lys245-Ser359 Accession # Q15583 | |
| 25 μg | |
| Primary | |
| Detects human TGIF1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 790707 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5'-TG-3' interacting factor, homeobox protein TGIF, homeobox protein TGIF1, HPE4, MGC39747, TALE homeobox TG-interacting factor, TGFB-induced factor (TALE family homeobox), TGFB-induced factor homeobox 1, TGIFMGC5066,5'-TG-3'-interacting factor 1, transforming growth factor-beta-induced factor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7050 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction