Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TGIF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7555
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7555 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7555 Supplier R&D Systems Supplier No. AF7555
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

TGIF1 Polyclonal specifically detects TGIF1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TGIF1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. Q15583
Gene Alias 5'-TG-3' interacting factor, homeobox protein TGIF, homeobox protein TGIF1, HPE4, MGC39747, TALE homeobox TG-interacting factor, TGFB-induced factor (TALE family homeobox), TGFB-induced factor homeobox 1, TGIFMGC5066,5'-TG-3'-interacting factor 1, transforming growth factor-beta-induced factor
Gene Symbols TGIF1
Host Species Sheep
Immunogen E. coli-derived recombinant human TGIF1 Lys245-Ser359 Accession # Q15583
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7050
Test Specificity Detects human TGIF1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.