Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Themis Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC6929R100UG
This item is not returnable.
View return policy
Description
Themis Monoclonal specifically detects Themis in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Themis | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8N1K5 | |
THEMIS | |
E. coli-derived recombinant human Themis Ala2-Pro282 Accession # Q8N1K5 | |
RUO | |
387357 | |
Human | |
IgG2a |
Flow Cytometry | |
701010 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, SPOT, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
Mouse | |
100 μg | |
Primary | |
Detects human Themis in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Themis is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction