Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thimet Oligopeptidase/THOP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | Thimet Oligopeptidase/THOP1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Polyclonal |
Description
Thimet Oligopeptidase/THOP1 Polyclonal specifically detects Thimet Oligopeptidase/THOP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Thimet Oligopeptidase/THOP1 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
EC 3.4.24, EC 3.4.24.15, Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, thimet oligopeptidase, thimet oligopeptidase 1, TOP | |
THOP1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
RUO | |
P52888 | |
7064 | |
E. coli-derived recombinant human Oligopeptidase/THOP1 Lys2-Cys689 Accession # P52888 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Oligopeptidase/THOP1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human Neurolysin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title