Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thioredoxin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.50
Specifications
Antigen | Thioredoxin-1 |
---|---|
Clone | 763629 |
Dilution | Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1970SP
![]() |
R&D Systems
MAB1970SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1970100
![]() |
R&D Systems
MAB1970100 |
100 μg |
Each for $444.50
|
|
|||||
Description
Thioredoxin-1 Monoclonal specifically detects Thioredoxin-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Thioredoxin-1 | |
Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
7295.0 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
763629 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Human | |
P10599 | |
E. coli-derived recombinant human Thioredoxin-1, Met1-Val105, Accession No. P10599 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Thioredoxin-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title