Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human THOC2 Partial ORF (NP_065182.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101508
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

THO2 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), HPR1 (MIM 606930), ALY (MIM 604171), and UAP56 (MIM 606390).[supplied by OMIM]

Sequence: MFVSDTVLKERLDPETLESLGLIKQSQQFNQKSVKIKTKLFYKQQKFNLLREENEGYAKLIAELGQDLSGSITSDLILENIKSLIGCFNLDPNRVLDVI

Specifications

Accession Number NP_065182.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57187
Molecular Weight (g/mol) 36.63kDa
Name THOC2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MFVSDTVLKERLDPETLESLGLIKQSQQFNQKSVKIKTKLFYKQQKFNLLREENEGYAKLIAELGQDLSGSITSDLILENIKSLIGCFNLDPNRVLDVI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CXorf3/THO2/dJ506G2.1
Common Name THOC2
Gene Symbol THOC2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.