Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human THRAP4 Partial ORF (AAH11375, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964699
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MKVVNLKQAILQAWKERWSDYQWAINMKKFFPKGATWDILNLADALLEQAMIGPSPNPLILSYLKYAISSQMVSYSSVLTAISKFDDFSRDLCVQALLDIMDMFCDRLSC

Specifications

Accession Number AAH11375
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9862
Molecular Weight (g/mol) 37.84kDa
Name THRAP4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MKVVNLKQAILQAWKERWSDYQWAINMKKFFPKGATWDILNLADALLEQAMIGPSPNPLILSYLKYAISSQMVSYSSVLTAISKFDDFSRDLCVQALLDIMDMFCDRLSC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ARC100/CRSP100/CRSP4/DRIP100/KIAA0130/MGC8748/THRAP4/TRAP100
Common Name MED24
Gene Symbol MED24
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.