Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Thrombomodulin/BDCA-3 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3947A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3947A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3947A Supplier R&D Systems Supplier No. FAB3947A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Thrombomodulin/BDCA-3
Applications Flow Cytometry
Classification Monoclonal
Clone 501733
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P07204(Val473)
Gene Alias BDCA3, BDCA-3, CD141, CD141 antigen, fetomodulin, THBD, THRM, thrombomodulin, TM
Gene Symbols THBD
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Thrombomodulin/BDCA-3 Ala19-Ser515 Accession # P07204(Val473)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7056
Test Specificity Detects human Thrombomodulin/BDCA-3 in ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.