Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thrombopoietin R/Tpo R PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1016P
This item is not returnable.
View return policy
Description
Thrombopoietin R/Tpo R Monoclonal specifically detects Thrombopoietin R/Tpo R in Human samples. It is validated for Flow Cytometry.Specifications
Thrombopoietin R/Tpo R | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD110, CD110 antigen, C-MPL, MPL, MPLV, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TpoR, TPO-R, TPORMPLV | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4352 | |
Human | |
Purified |
Flow Cytometry | |
167639 | |
Flow Cytometry 10 uL/10^6 cells | |
P40238 | |
MPL | |
Mouse myeloma cell line NS0-derived recombinant human Thrombopoietin R/Tpo R Ser25-Tyr423 Accession # P40238 | |
100 Tests | |
Primary | |
Detects human Thrombopoietin R/Tpo R in Western blots. In Western blots, no cross-reactivity with recombinant mouse Tpo R or recombinant human Epo R is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction