Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Thrombopoietin/Tpo Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB288500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB288500 500 μg
MAB288SP 25 μg
MAB288100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB288500 Supplier R&D Systems Supplier No. MAB288500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thrombopoietin/THPO Monoclonal specifically detects Thrombopoietin/THPO in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Thrombopoietin/Tpo
Applications Neutralization, ELISA
Classification Monoclonal
Clone 34817
Conjugate Unconjugated
Dilution Neutralization 5-10 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P40225
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Gene Symbols THPO
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Thrombopoietin/Tpo Ser22-Gly353 Accession # P40225
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7066
Test Specificity Detects human Thrombopoietin/Tpo in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) G-CSF, rhG-CSF R, rhGM-CSF, recombinant mouse GM-CSF, or rhM-CSF is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.