Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thrombospondin-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF3074
This item is not returnable.
View return policy
Description
Thrombospondin-1 Polyclonal specifically detects Thrombospondin-1 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
Thrombospondin-1 | |
Polyclonal | |
Biotin | |
P07996 | |
THBS1 | |
Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-1 Asn19-Phe1170 Accession # P07996 | |
50 μg | |
Primary | |
Detects Thrombospondin-1 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) Thrombospondin-2 and rhThrombospondin-4 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
THBS, THBS1, THBS-1, thrombospondin 1, Thrombospondin1, thrombospondin-1p180, TSP-1, TSP1thrombospondin-1, TSPTSP-1 | |
Goat | |
Affinity Purified | |
RUO | |
7057 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction