Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thrombospondin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB16351500
This item is not returnable.
View return policy
Description
Thrombospondin-2 Monoclonal specifically detects Thrombospondin-2 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Thrombospondin-2 | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P35442 | |
| THBS2 | |
| Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-2 Gly19-Ile1172 Accession # P35442 | |
| 500 μg | |
| Primary | |
| Detects human Thrombospondin-2 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Thrombospondin-1, -4, rhMMP-2, or rhCD36 is observed. | |
| Human | |
| Supernatant |
| ELISA | |
| 230927 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| THBS2, thrombospondin 2, Thrombospondin2, TSP-2, TSP2thrombospondin-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7058 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction