Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thrombospondin-2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1635
This item is not returnable.
View return policy
Description
Thrombospondin-2 Polyclonal specifically detects Thrombospondin-2 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
Thrombospondin-2 | |
Polyclonal | |
Biotin | |
P35442 | |
THBS2 | |
Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-2 Gly19-Ile1172 Accession # P35442 | |
50 μg | |
Primary | |
Detects human Thrombospondin-2 in ELISAs and Western blots. In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant human (rh) THBS-1 and rhTHBS-4 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
THBS2, thrombospondin 2, Thrombospondin2, TSP-2, TSP2thrombospondin-2 | |
Goat | |
Affinity Purified | |
RUO | |
7058 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction