Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Thrombospondin-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2390SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2390SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2390SP Supplier R&D Systems Supplier No. AF2390SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 7 publications

Thrombospondin-4 Polyclonal specifically detects Thrombospondin-4 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Thrombospondin-4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P35443
Gene Alias THBS4, thrombospondin 4, thrombospondin-4, Thrombospondin4, TSP4, TSP-4
Gene Symbols THBS4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-4 Ala22-Asn961 (Pro276Ala, Ala420Val) Accession # P35443
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7060
Test Specificity Detects human Thrombospondin-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Thrombospondin-4 is observed and less than 5% cross-reactivity with recombinant human (rh) Thrombospondin-1, rhThrombospondin-2, and rhThrombospondin-3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.