Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human THRSP Partial ORF (NP_003242.1, 1 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964709 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-709 25 ug
89-964-708 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-709 Supplier Abnova™ Supplier No. H00007069Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is similar to the gene product of S14, a rat gene whose expression is limited to liver and adipose tissue and is controlled by nutritional and hormonal factors. This gene has been shown to be expressed in liver and adipocytes, particularly in lipomatous modules. It is also found to be expressed in lipogenic breast cancers, which suggests a role in controlling tumor lipid metabolism. [provided by RefSeq]

Sequence: MQVLTKRYPKNCLLTVMDRYAAEVHNMEQVVMIPSLLRDVQLSGPGGQAQAEAPDLYTYFTMLKAICVDVDHGLLPREEWQAKV

Specifications

Accession Number NP_003242.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7069
Molecular Weight (g/mol) 34.98kDa
Name THRSP (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MQVLTKRYPKNCLLTVMDRYAAEVHNMEQVVMIPSLLRDVQLSGPGGQAQAEAPDLYTYFTMLKAICVDVDHGLLPREEWQAKV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC21659/S14/SPOT14
Common Name THRSP
Gene Symbol THRSP
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.