Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human THSD1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5178T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5178T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5178T100 Supplier R&D Systems Supplier No. FAB5178T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

THSD1 Monoclonal specifically detects THSD1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen THSD1
Applications Flow Cytometry
Classification Monoclonal
Clone 541213
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_954872
Gene Alias thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSP, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010
Gene Symbols THSD1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human THSD1 isoform 2 Glu25-Ile361 Accession # NP_954872
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55901
Test Specificity Detects human TSHD1.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.