Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human THSD1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5178R100UG
This item is not returnable.
View return policy
Description
THSD1 Monoclonal specifically detects THSD1 in Human samples. It is validated for Flow Cytometry.Specifications
THSD1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_954872 | |
THSD1 | |
Mouse myeloma cell line NS0-derived recombinant human THSD1 isoform 2 Glu25-Ile361 Accession # NP_954872 | |
RUO | |
55901 | |
Human | |
IgG1 |
Flow Cytometry | |
541213 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSP, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010 | |
Mouse | |
100 μg | |
Primary | |
Detects human TSHD1. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction