Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human THSD1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5178
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5178 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5178 Supplier R&D Systems Supplier No. MAB5178
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

THSD1 Monoclonal specifically detects THSD1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen THSD1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 541213
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_954872
Gene Alias thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSP, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010
Gene Symbols THSD1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human THSD1 isoform 2 Glu25-Ile361 Accession # NP_954872
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55901
Test Specificity Detects human TSHD1.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.