Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thymidine Kinase 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | Thymidine Kinase 1 |
---|---|
Clone | 1008515 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8180SP
![]() |
R&D Systems
MAB8180SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8180100
![]() |
R&D Systems
MAB8180100 |
100 μg |
Each for $432.50
|
|
|||||
Description
Thymidine Kinase 1 Monoclonal specifically detects Thymidine Kinase 1 in Human samples. It is validated for Immunocytochemistry.Specifications
Thymidine Kinase 1 | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK1, TK2 | |
E. coli-derived recombinant human Thymidine Kinase 1, Met1-Asn234, Accession # P04183 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Thymidine Kinase 1 in direct ELISAs. |
1008515 | |
Immunocytochemistry | |
Unconjugated | |
Human | |
P04183 | |
7083 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title