Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Thymidine Kinase 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8180SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8180SP 25 μg
AF8180100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF8180SP Supplier R&D Systems Supplier No. AF8180SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Antibody

Thymidine Kinase 1 specifically detects Thymidine Kinase 1 in Human samples. It is validated for Simple Western, Immunohistochemistry.

Specifications

Antigen Thymidine Kinase 1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Simple Western 20 ug/mL, Immunohistochemistry 1-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P04183
Gene Alias EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK1, TK2
Host Species Sheep
Immunogen E. coli-derived human Thymidine Kinase 1 protein, Ser2-Asn234, Accession No. P04183
Purification Method Antigen Affinity-purified
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 7083.0
Test Specificity Detects human Thymidine Kinase 1 Protein in direct ELISAs.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.