Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thymopoietin Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Thymopoietin/LAP2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Thymopoietin/LAP2 Polyclonal specifically detects Thymopoietin/LAP2 in Human samples. It is validated for Western Blot.Specifications
Thymopoietin/LAP2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
7112 | |
E. coli-derived recombinant human Thymopoietin Pro2-Gln187 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Thymopoietin in direct ELISAs and Western blots. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
CMD1T, lamina-associated polypeptide 2, LAP2PRO0868, LEM domain containing 4, LEMD4, MGC61508, thymopoietin, Thymopoietin isoform alpha, Thymopoietin, isoforms beta/gamma, Thymopoietin-related peptide isoform alpha, Thymopoietin-related peptide isoforms beta/gamma, TMPO, TP, TP alpha, TP beta/gamma, TPRP isoform alpha, TPRP isoforms beta/gamma | |
TMPO | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title