Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Tie-2 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP183746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP183746 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP183746 Supplier Novus Biologicals™ Supplier No. NBP183746

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human Tie-2 ELISA Kit (Colorimetric) measures human Tie-2 in cell culture supernatant, serum, plasma (edta, citrate, heparin).

Specifications

Assay Range 0.16 - 10.0ng/mL
Assay Sensitivity 0.03ng/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity The assay detects both natural and recombinant human TIE-2. The interference of circulating factors of the immune systeme was evaluated by spiking these proteins at physiologically relevant concentrations into serum. There was no crossreactivity detected, notably not with TIE-1.
For Use With (Application) ELISA
Gene ID (Entrez) 7010
Gene Symbol TEK
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human TIE-2, Biotin-Conjugate anti-human TIE-2 polyclonal antibody, Streptavidin-HRP, Human TIE-2 Standard lyophilized, 20ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Angiogenesis, Cancer, Protein Kinase
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase
Target Tie-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.