Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Tie-2 Quantikine ELISA Kit
Click to view available options
Sufficient For:
1 x 96-well Plate
Description
The Human Tie-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Tie-2 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Specifications
Specifications
Assay Range | 0.16 - 10ng/mL |
Assay Sensitivity | 0.066ng/mL |
Conjugate | HRP |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
Specificity | Natural and Recombinant Human Tie-2 |
Sufficient For | 1 x 96-well Plate |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 7010 |
Sample Volume | Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL) |
Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction