Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TIGIT Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $596.00
Specifications
Antigen | TIGIT |
---|---|
Clone | 741170 |
Dilution | Blockade of Receptor-ligand Interaction 70-350 ng/mL |
Applications | Blockade of Receptor-ligand Interaction |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB78981SP
![]() |
R&D Systems
MAB78981SP |
25 μg |
Each for $214.00
|
|
|||||
MAB78981100
![]() |
R&D Systems
MAB78981100 |
100 μg |
Each for $596.00
|
|
|||||
Description
TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
TIGIT | |
Blockade of Receptor-ligand Interaction 70-350 ng/mL | |
Monoclonal | |
Mouse | |
Human | |
Q495A1 | |
Mouse myeloma cell line NS0-derived recombinant human TIGIT Met1-Pro141 Accession # Q495A1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TIGIT in direct ELISAs. |
741170 | |
Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
201633 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title