Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Tiki2/TRABD2B Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | Tiki2/TRABD2B |
---|---|
Dilution | Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Tiki2/TRABD2B Polyclonal specifically detects Tiki2/TRABD2B in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Tiki2/TRABD2B | |
Western Blot, Flow Cytometry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
HKAT, TIKI2, TRABD2B, TRABD2B TraB domain containing 2B | |
Human embryonic kidney cell line HEK293-derived transfected with human Tiki2/TRABD2B Accession # A6NFA1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Tiki2/TRABD2B in direct ELISAs and Western blots. |
Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
A6NFA1 | |
388630 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title