Learn More
Human TIM-1/KIM-1/HAVCR Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | TIM-1/KIM-1/HAVCR |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 990605 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide |
| Gene Accession No. | Q96D42 |
| Gene Alias | CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.