Learn More
Human TIM-1/KIM-1/HAVCR Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB17503S100UG
Catalog No. FB17503S100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TIM-1/KIM-1/HAVCR
Monoclonal
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Human embryonic kidney cell, HEK293-derived human TIM-1/KIM-1/HAVCR, Met1-Thr288, Accession No. Q96D42
100 μg
26762.0
Human
Supernatant
Flow Cytometry
990605
Flow Cytometry 0.25-1 ug/10^6 cells
Q96D42
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human TIM-1/KIM-1/HAVCR in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.