Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TIM-3 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB2365RT100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2365RT100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB2365RT100 Supplier R&D Systems Supplier No. FAB2365RT100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TIM-3
Applications Flow Cytometry
Classification Monoclonal
Clone 344823R
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1μg/106 cells
Formulation Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8TDQ0.2
Gene Alias CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIM-3 Ser22-Arg200 Accession # Q8TDQ0.2
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 84868
Test Specificity Detects human TIM-3 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.