Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TIM-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB23651
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB23651 100 μg
MAB23651SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB23651 Supplier R&D Systems Supplier No. MAB23651
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Agonist Activity.

Specifications

Applications Agonist Activity
Classification Monoclonal
Clone 344801
Concentration LYOPH
Conjugate Unconjugated
Dilution Agonist Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8TDQ0
Gene Alias CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIM-3 Met1-Arg200 Accession # Q8TDQ0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Test Specificity Detects recombinant human TIM-3 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.