Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TIM-3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody has been used in 3 publications
$227.50 - $503.00
Specifications
Antigen | TIM-3 |
---|---|
Clone | 2321C |
Dilution | Western Blot 2 μg;/mL, Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg;/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB23652SP
![]() |
R&D Systems
MAB23652SP |
25 μg |
Each for $227.50
|
|
|||||
MAB23652100
![]() |
R&D Systems
MAB23652100 |
100 μg |
Each for $503.00
|
|
|||||
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
TIM-3 | |
Western Blot 2 μg;/mL, Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg;/mL, CyTOF-ready | |
Monoclonal | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
Mouse myeloma cell line NS0-derived recombinant human TIM-3, Met1-Arg200, Accession No. Q8TDQ0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TIM-3 in direct ELISAs. |
2321C | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Human | |
Q8TDQ0 | |
84868.0 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title